N-Acetyl-D-Glucosamin, ≥98 %
N-Acetyl-D-Glucosamin, ≥98 %
N-Acetyl-D-Glucosamin, ≥98 %
N-Acetyl-D-Glucosamin, ≥98 %
Thermo Scientific Chemicals

N-Acetyl-D-Glucosamin, ≥98 %

N-Acetyl-D-glucosamin, CAS-Nr. 72-87-7, ist das D-Isomer des aus Glucose gewonnenen Monomers, das in strukturellen Polysacchariden gefunden wird.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
50 g
100 g
Katalognummer A13047.18
auch als A13047-18 bezeichnet
Preis (EUR)
-
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS7512-17-6
IUPAC NameN-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular FormulaC8H15NO6
InChI KeyOVRNDRQMDRJTHS-WZWLWIOANA-N
SMILESCC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥98.0% (HPLC)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
Mehr anzeigen
N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Löslich in konzentrierter Salzsäure, Schwefelsäure, Phosphorsäure und Ameisensäure. Unlöslich in Wasser, verdünnten Säuren, verdünnten Alkalien, konzentrierten Alkalien und organischen Lösungsmittel.

Hinweise
Hygroskopisch. Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
RUO – Research Use Only

Allgemeine Referenzen:

  1. Bobbink, F. D.; Zhang, J.; Pierson, Y.; Chen, X.; Yan, N. Conversion of chitin derived N-acetyl-D-glucosamine (NAG) into polyols over transition metal catalysts and hydrogen in water. Green Chem. 2015, 17 (2), 1024-1031.
  2. Mutter, S. T.; Zielinski, F.; Cheeseman, J. R.; Johannessen, C.; Popelier, P. L.; Blanch, E. W. Conformational dynamics of carbohydrates: Raman optical activity of D-glucuronic acid and N-acetyl-D-glucosamine using a combined molecular dynamics and quantum chemical approach. Phys. Chem. Chem. Phys. 2015, 17 (8), 6016-6027.