Thermo Scientific Chemicals

4-Chlorophenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13065.36
500 g, Each
Thermo Scientific Chemicals

4-Chlorophenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13065.36
500 g, Each
Quantity
Catalog number: A13065.36
also known as A13065-36
Price (USD)
Price: 104.00
Online price: 89.65
Your price:
Quantity
-

Chemical Identifiers

CAS
122-88-3
IUPAC Name
2-(4-chlorophenoxy)acetic acid
Molecular Formula
C8H7ClO3
InChI Key
SODPIMGUZLOIPE-UHFFFAOYSA-N
SMILES
OC(=O)COC1=CC=C(Cl)C=C1
Identification (FTIR)
Conforms
Melting Point (clear melt)
155.0-161.0?C
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text