Thermo Scientific Chemicals

5-Nitro-1H-indazole, 98+%, Thermo Scientific Chemicals

Catalog number: A13478.14
25 g, Each
Thermo Scientific Chemicals

5-Nitro-1H-indazole, 98+%, Thermo Scientific Chemicals

Catalog number: A13478.14
25 g, Each
Quantity
Catalog number: A13478.14
also known as A13478-14
Price (USD)
Price: 83.00
Online price: 71.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5401-94-5
IUPAC Name
5-nitro-1H-indazole
Molecular Formula
C7H5N3O2
InChI Key
WSGURAYTCUVDQL-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C2NN=CC2=C1
Appearance (Color)
White to cream or yellow to brown
Form
Crystals or powder or crystalline powder and/or lumpy powder
Melting Point (clear melt)
206.5-212.5?C
Assay (HPLC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text