P-Tolylisocyanat, 99 %, Thermo Scientific Chemicals
P-Tolylisocyanat, 99 %, Thermo Scientific Chemicals
P-Tolylisocyanat, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

P-Tolylisocyanat, 99 %, Thermo Scientific Chemicals

CAS: 622-58-2 | C8H7NO | 133.15 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
50 g
100 g
Katalognummer A13577.18
auch als A13577-18 bezeichnet
Preis (EUR)
147,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
147,00
Each
Chemikalien-Kennzeichnungen
CAS622-58-2
IUPAC Name1-isocyanato-4-methylbenzene
Molecular FormulaC8H7NO
InChI KeyMGYGFNQQGAQEON-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)N=C=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.5300-1.5330 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only