Diethyliodomethylphosphonat, 98+%, Thermo Scientific Chemicals
Diethyliodomethylphosphonat, 98+%, Thermo Scientific Chemicals
Diethyliodomethylphosphonat, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyliodomethylphosphonat, 98+%, Thermo Scientific Chemicals

CAS: 10419-77-9 | C5H12IO3P | 278.03 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer A13697.06
auch als A13697-06 bezeichnet
Preis (EUR)
78,70
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
78,70
Each
Chemikalien-Kennzeichnungen
CAS10419-77-9
SpecificationsSpecification SheetDatenblatt
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4970-1.5010 @ 20?C
Assay (GC)≥98.0%
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Diethyl-(iodmethyl)phosphonat wird in der Synthese von H2N-Tyr(tBu)-Vinylsulfon verwendet.

Löslichkeit
Nicht mit Wasser mischbar.

Hinweise
Lichtempfindlich. Nicht kompatibel mit starken Oxidationsmitteln.
RUO – Research Use Only

Allgemeine Referenzen:

  1. Linden, W. A.; Segal, E.; Child, M. A.; Byzia, A.; Drąg, M.; Bogyo, M. Design and Synthesis of Activity-Based Probes and Inhibitors for Bleomycin Hydrolase. Chem. Biol. 2015, 22 (8), 995-1001.
  2. Li, Y.; Huang, H.; Wang, Z.; Yang, F.; Li, D.; Qin, B.; Ren, X. Stereoselective synthesis of C-sulfonylated aziridines from halomethyl phenyl sulfone and N-tert-butanesulfinyl imines. RSC Adv. 2014, 4 (2), 969-973.