DL-Tyrosine, 98%
Thermo Scientific Chemicals

DL-Tyrosine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13740.09
also known as A13740-09
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.20 
Save 4.55 (14%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.20 
Save 4.55 (14%)
Add to cart
DL-Tyrosine, 98%
Catalog numberA13740.09
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.20 
Save 4.55 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyOUYCCCASQSFEME-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(O)C=C1)C(O)=O
Molecular Weight (g/mol)181.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
FormPowder
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Melvin Fields,; Doris E. Walz,; Seymour Rothchild. The Synthesis of Ethyl Acetamidocyanoacetate, DL-Lysine, DL-Ornithine and DL-Tyrosine Labeled with Isotopic Carbon1. J. Am. Chem. Soc., 1951, 73(3),1000-1002.
  2. Yoshie Inomata,; Tadaaki Inomata,; Takao Moriwaki. Infrared Absorption Spectra and Normal Coordinate Analysis of Metal-DL-Tyrosine Chelates. Bulletin of the Chemical Society of Japan. 1974 , 47 (4), 818-824.