Thermo Scientific Chemicals

DL-Tyrosine, 98%

Catalog number: A13740.30
250 g, Each
Thermo Scientific Chemicals

DL-Tyrosine, 98%

Catalog number: A13740.30
250 g, Each
Quantity
Catalog number: A13740.30
also known as A13740-30
Price (USD)

Chemical Identifiers

CAS
556-03-6
IUPAC Name
2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C9H11NO3
InChI Key
OUYCCCASQSFEME-UHFFFAOYNA-N
SMILES
NC(CC1=CC=C(O)C=C1)C(O)=O
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥97.0 to ≤103.0%
Form
Powder

Description

The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text