Thermo Scientific Chemicals

Hexanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13789.AE
100 mL, Each
Thermo Scientific Chemicals

Hexanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13789.AE
100 mL, Each
Quantity
Catalog number: A13789.AE
also known as A13789-AE
Price (USD)
Price: 25.10
Online price: 21.65
Your price:
Quantity
-

Chemical Identifiers

CAS
142-62-1
IUPAC Name
hexanoic acid
Molecular Formula
C6H12O2
InChI Key
FUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILES
CCCCCC(O)=O
Assay (Silylated GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

Solubility
Miscible with diethyl ether and ethanol. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and allyl alcohol.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text