Benzylphosphonsäure, 97 %, Thermo Scientific Chemicals
Benzylphosphonsäure, 97 %, Thermo Scientific Chemicals
Benzylphosphonsäure, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzylphosphonsäure, 97 %, Thermo Scientific Chemicals

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
Katalognummer A13850.03
auch als A13850-03 bezeichnet
Preis (EUR)
31,60
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
31,60
Each
Chemikalien-Kennzeichnungen
CAS6881-57-8
SpecificationsSpecification SheetDatenblatt
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream
FormPowder
Melting Point (clear melt)165.5-174.5?C
Structural mimic of phosphotyrosine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only