Thermo Scientific Chemicals

Thiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13941.36
500 g, Each
Thermo Scientific Chemicals

Thiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13941.36
500 g, Each
Quantity
Catalog number: A13941.36
also known as A13941-36
Price (USD)
Price: 63.30
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-02-1
IUPAC Name
thiophene
Molecular Formula
C4H4S
InChI Key
YTPLMLYBLZKORZ-UHFFFAOYSA-N
SMILES
S1C=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.5265-1.5295 @ 20°C

Description

Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text