Thermo Scientific Chemicals

Methyl 2-furoate, 98+%, Thermo Scientific Chemicals

Catalog number: A14009.36
500 g, Each
Thermo Scientific Chemicals

Methyl 2-furoate, 98+%, Thermo Scientific Chemicals

Catalog number: A14009.36
500 g, Each
Quantity
Catalog number: A14009.36
also known as A14009-36
Price (USD)
Price: 296.00
Online price: 258.65
Your price:
Quantity
-

Chemical Identifiers

CAS
611-13-2
IUPAC Name
methyl furan-2-carboxylate
Molecular Formula
C6H6O3
InChI Key
HDJLSECJEQSPKW-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CO1
Assay (GC)
≥98.0%
Refractive Index
1.4830-1.4900 @ 20°C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text