Thermo Scientific Chemicals

Lepidine, 97%, Thermo Scientific Chemicals

Catalog number: A14040.30
250 g, Each
Thermo Scientific Chemicals

Lepidine, 97%, Thermo Scientific Chemicals

Catalog number: A14040.30
250 g, Each
Quantity
Catalog number: A14040.30
also known as A14040-30
Price (USD)
Price: 807.00
Online price: 704.65
Your price:
Quantity
-

Chemical Identifiers

CAS
491-35-0
IUPAC Name
4-methylquinoline
Molecular Formula
C10H9N
InChI Key
MUDSDYNRBDKLGK-UHFFFAOYSA-N
SMILES
CC1=C2C=CC=CC2=NC=C1
Refractive Index
1.6155-1.6205 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow

Description

Lepidine is used in the preparation of certain dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lepidine is used in the preparation of certain dyes.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text