Thermo Scientific™

m-Toluidine, 99%, Thermo Scientific Chemicals

Catalog number: A14058.0B
1000 g, Each
Thermo Scientific™

m-Toluidine, 99%, Thermo Scientific Chemicals

Catalog number: A14058.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14058.0B
also known as A14058-0B
Price (USD)
Price: 104.00
Online price: 99.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-44-1
IUPAC Name
3-methylaniline
Molecular Formula
C7H9N
InChI Key
JJYPMNFTHPTTDI-UHFFFAOYSA-N
SMILES
CC1=CC=CC(N)=C1
Comment
May darken on storage
Refractive Index
1.5655-1.5685 @ 20?C
Appearance (Color)
Clear colorless to yellow or brown
Form
Liquid
Assay (GC)
≥98.5%

Description

m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

Solubility
Soluble in water (2g/L), (10 g/L at 25°C ).

Notes
Keep container tightly sealed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text