Thermo Scientific Chemicals

4-Aminothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A14082.14
25 g, Each
Thermo Scientific Chemicals

4-Aminothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A14082.14
25 g, Each
Quantity
Catalog number: A14082.14
also known as A14082-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1193-02-8
IUPAC Name
(4-aminophenyl)sulfanide
Molecular Formula
C6H6NS
InChI Key
WCDSVWRUXWCYFN-UHFFFAOYSA-M
SMILES
NC1=CC=C([S-])C=C1
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
33.0-43.0?C
Appearance (Color)
Yellow
Form
Fused solid

Description

4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text