Thermo Scientific Chemicals

4-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A14130.06
5 g, Each
Thermo Scientific Chemicals

4-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A14130.06
5 g, Each
Quantity
Catalog number: A14130.06
also known as A14130-06
Price (USD)
Price: 110.00
Online price: 93.65
Your price:
Quantity
-

Chemical Identifiers

CAS
623-04-1
IUPAC Name
(4-aminophenyl)methanol
Molecular Formula
C7H9NO
InChI Key
AXKGIPZJYUNAIW-UHFFFAOYSA-N
SMILES
NC1=CC=C(CO)C=C1
Comment
Specification differs for U.S. and non-U.S. material where indicated
Assay (GC)
≥97.5% (non-U.S. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. specification)
Melting Point (clear melt)
56-65°C

Description

4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

Solubility
Soluble in alcohol, ether and benzene. Partially soluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents and strong acids. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text