Thermo Scientific Chemicals

2-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14410.30
250 g, Each
Thermo Scientific Chemicals

2-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14410.30
250 g, Each
Quantity
Catalog number: A14410.30
also known as A14410-30
Price (USD)
Price: 228.00
Online price: 193.65
Your price:
Quantity
-

Chemical Identifiers

CAS
90-01-7
IUPAC Name
2-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
CQRYARSYNCAZFO-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=C1O
Appearance (Color)
Pale cream to cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
80.0-87.0?C
Assay (GC)
≥98.5%

Description

2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text