Thermo Scientific Chemicals

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A14472.22
100 g, Each
Thermo Scientific Chemicals

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A14472.22
100 g, Each
Quantity
Catalog number: A14472.22
also known as A14472-22
Price (USD)
Price: 492.00
Online price: 418.65
Your price:
Quantity
-

Chemical Identifiers

CAS
356-42-3
IUPAC Name
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
Molecular Formula
C6F10O3
InChI Key
XETRHNFRKCNWAJ-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)F
Form
Liquid
Refractive Index
<1.3000 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%

Description

Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

Solubility
Reacts with water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text