Thermo Scientific Chemicals

o-Tolylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A14787.14
25 g, Each
Thermo Scientific Chemicals

o-Tolylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A14787.14
25 g, Each
Quantity
Catalog number: A14787.14
also known as A14787-14
Price (USD)
Price: 81.90
Online price: 70.65
Your price:
Quantity
-

Chemical Identifiers

CAS
22364-68-7
IUPAC Name
2-(2-methylphenyl)acetonitrile
Molecular Formula
C9H9N
InChI Key
WMGVPDQNPUQRND-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1CC#N
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Refractive Index
1.5255-1.5295 @ 20?C
Form
Liquid

Description

o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

Solubility
Slightly soluble in water. Soluble in benzene, ethyl ether, ethanol.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text