Thermo Scientific Chemicals

Tri-n-butylamine, 98%, Thermo Scientific Chemicals

Catalog number: A14868.0F
2500 mL, Each
Thermo Scientific Chemicals

Tri-n-butylamine, 98%, Thermo Scientific Chemicals

Catalog number: A14868.0F
2500 mL, Each
Quantity
Catalog number: A14868.0F
also known as A14868-0F
Price (USD)

Chemical Identifiers

CAS
102-82-9
IUPAC Name
tributylamine
Molecular Formula
C12H27N
InChI Key
IMFACGCPASFAPR-UHFFFAOYSA-N
SMILES
CCCCN(CCCC)CCCC
Assay from Supplier's CofA
≥97.5% (U.S. sourced material)
Refractive Index
1.4270-1.4310 @ 20°C (non-U.S. sourced material)
Assay (GC)
≥97.5% (non-U.S. sourced material)
Identification (FTIR)
Conforms (non-U.S. sourced material)
Appearance (Color)
Clear colorless to pale yellow

Description

Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text