Thermo Scientific Chemicals

4-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14993.03
1 g, Each
Thermo Scientific Chemicals

4-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14993.03
1 g, Each
Quantity
Catalog number: A14993.03
also known as A14993-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
611-95-0
IUPAC Name
4-benzoylbenzoic acid
Molecular Formula
C14H10O3
InChI Key
IFQUPKAISSPFTE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Powder or crystalline powder
Assay (Silylated GC)
≥98.5%

Description

4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
It is soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text