Thermo Scientific Chemicals

Malononitrile, 99%, Thermo Scientific Chemicals

Catalog number: A15046.36
500 g, Each
Thermo Scientific Chemicals

Malononitrile, 99%, Thermo Scientific Chemicals

Catalog number: A15046.36
500 g, Each
Quantity
Catalog number: A15046.36
also known as A15046-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
109-77-3
IUPAC Name
propanedinitrile
Molecular Formula
C3H2N2
InChI Key
CUONGYYJJVDODC-UHFFFAOYSA-N
SMILES
N#CCC#N
Appearance (Color)
Colorless to white to pale cream to cream to yellow to orange to brown
Form
Crystals or powder or crystalline powder or fused solid or clear liquid as melt
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
29.0-35.0?C

Description

Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text