Thermo Scientific Chemicals

2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A15066.36
500 g, Each
Thermo Scientific Chemicals

2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A15066.36
500 g, Each
Quantity
Catalog number: A15066.36
also known as A15066-36
Price (USD)
Price: 194.00
Online price: 164.65
Your price:
Quantity
-

Chemical Identifiers

CAS
75-30-9
IUPAC Name
2-iodopropane
Molecular Formula
C3H7I
InChI Key
FMKOJHQHASLBPH-UHFFFAOYSA-N
SMILES
CC(C)I
Form
Liquid
Refractive Index
1.4960-1.5000 @ 20?C
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Stabilizer
Copper powder

Description

2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text