Thermo Scientific Chemicals

Tri-n-octylamine, 95%, Thermo Scientific Chemicals

Catalog number: A15067.AP
500 mL, Each
Thermo Scientific Chemicals

Tri-n-octylamine, 95%, Thermo Scientific Chemicals

Catalog number: A15067.AP
500 mL, Each
Quantity
Catalog number: A15067.AP
also known as A15067-AP
Price (USD)
Price: 234.00
Online price: 198.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1116-76-3
IUPAC Name
trioctylamine
Molecular Formula
C24H51N
InChI Key
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
SMILES
CCCCCCCCN(CCCCCCCC)CCCCCCCC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Refractive Index
1.4465-1.4525 @ 20?C

Description

Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text