Thermo Scientific Chemicals

1,3-Propanedithiol, 97%, Thermo Scientific Chemicals

Catalog number: A15261.06
5 g, Each
Thermo Scientific Chemicals

1,3-Propanedithiol, 97%, Thermo Scientific Chemicals

Catalog number: A15261.06
5 g, Each
Quantity
Catalog number: A15261.06
also known as A15261-06
Price (USD)
Price: 26.60
Online price: 22.65
Your price:
Quantity
-

Chemical Identifiers

CAS
109-80-8
IUPAC Name
propane-1,3-dithiol
Molecular Formula
C3H8S2
InChI Key
ZJLMKPKYJBQJNH-UHFFFAOYSA-N
SMILES
SCCCS
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay (GC)
>96.0%
Refractive Index
1.5375-1.5425 @ 20?C

Description

1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

Solubility
Slightly soluble in water. Miscible with alcohol, ether, chloroform, etherMiscible with alcohol, ether, chloroform and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text