Thermo Scientific Chemicals

4-Carboxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15277.30
250 g, Each
Thermo Scientific Chemicals

4-Carboxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15277.30
250 g, Each
Quantity
Catalog number: A15277.30
also known as A15277-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
619-66-9
IUPAC Name
4-formylbenzoic acid
Molecular Formula
C8H6O3
InChI Key
GOUHYARYYWKXHS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=O)C=C1
Appearance (Color)
White to cream to pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

Solubility
Soluble in water, methanol, DMSO, ether, and chloroform.

Notes
Store away from air and strong bases. Air Sensitive. Incompatible with bases and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text