Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A15283.0C
10000 g, Each
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A15283.0C
10000 g, Each
Quantity
Catalog number: A15283.0C
also known as A15283-0C
Price (USD)
Price: 378.00
Online price: 321.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7722-76-1
IUPAC Name
phosphoric acid amine
Molecular Formula
H6NO4P
InChI Key
LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES
N.OP(O)(O)=O
Appearance (Color)
White
Form
Crystalline powder or crystals
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
197-206?C

Description

Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text