Thermo Scientific Chemicals

4-Bromo-3-methylaniline, 97%, Thermo Scientific Chemicals

Catalog number: A15417.18
50 g, Each
Thermo Scientific Chemicals

4-Bromo-3-methylaniline, 97%, Thermo Scientific Chemicals

Catalog number: A15417.18
50 g, Each
Quantity
Catalog number: A15417.18
also known as A15417-18
Price (USD)
Price: 86.10
Online price: 73.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6933-10-4
IUPAC Name
4-bromo-3-methylaniline
Molecular Formula
C7H8BrN
InChI Key
MMEGELSFOYDPQW-UHFFFAOYSA-N
SMILES
CC1=CC(N)=CC=C1Br
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Appearance (Color)
White to cream to pale brown or grey
Melting Point
76.0-84.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text