Thermo Scientific Chemicals

Cyanuric acid, 99%, Thermo Scientific Chemicals

Catalog number: A15447.22
100 g, Each
Thermo Scientific Chemicals

Cyanuric acid, 99%, Thermo Scientific Chemicals

Catalog number: A15447.22
100 g, Each
Quantity
Catalog number: A15447.22
also known as A15447-22
Price (USD)
Price: 44.90
Online price: 38.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-80-5
IUPAC Name
1,3,5-triazinane-2,4,6-trione
Molecular Formula
C3H3N3O3
InChI Key
ZFSLODLOARCGLH-UHFFFAOYSA-N
SMILES
O=C1NC(=O)NC(=O)N1
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5%
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Powder

Description

Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text