Thermo Scientific Chemicals

Isophthalaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15788.22
100 g, Each
Thermo Scientific Chemicals

Isophthalaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15788.22
100 g, Each
Quantity
Catalog number: A15788.22
also known as A15788-22
Price (USD)
Price: 608.00
Online price: 531.65
Your price:
Quantity
-

Chemical Identifiers

CAS
626-19-7
IUPAC Name
benzene-1,3-dicarbaldehyde
Molecular Formula
C8H6O2
InChI Key
IZALUMVGBVKPJD-UHFFFAOYSA-N
SMILES
O=CC1=CC(C=O)=CC=C1
Appearance (Color)
Pale cream to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Free acid (titration)
≤1%
Melting Point (clear melt)
85.0-91.0?C

Description

Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text