Thermo Scientific Chemicals

4-Methylmorpholine N-oxide monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15996.14
25 g, Each
Thermo Scientific Chemicals

4-Methylmorpholine N-oxide monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15996.14
25 g, Each
Quantity
Catalog number: A15996.14
also known as A15996-14
Price (USD)
Price: 74.40
Online price: 63.65
Your price:
Quantity
-

Chemical Identifiers

CAS
70187-32-5
IUPAC Name
4-methylmorpholin-4-ium-4-olate
Molecular Formula
C5H11NO2
InChI Key
LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES
C[N+]1([O-])CCOCC1
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (dry wt. basis)
Appearance (Color)
White to cream
Form
Crystalline powder and/or Lumps
Solution Test
Clear colorless (ethanol, 5%)
Water Content (Karl Fischer Titration)
10-16% (0.7-1.2 waters of hydration)

Description

4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text