Thermo Scientific Chemicals

Polyethylene glycol 4,000, Thermo Scientific Chemicals

Catalog number: A16151.30
250 g, Each
Thermo Scientific Chemicals

Polyethylene glycol 4,000, Thermo Scientific Chemicals

Catalog number: A16151.30
250 g, Each
Quantity
Catalog number: A16151.30
also known as A16151-30
Price (USD)
Price: 46.70
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)
Molecular Weight
3,600-4,400
Viscosity
110.0-158.0 cSt (U.S. Specification)
Form
Powder or flakes or granular

Description

Polyethylene glycol is used in gene delivery of protoplast transfection and incubation. It is used in the preparation of extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues. It is useful to mediate DNA transformation into protoplasts derived from rice leaf sheaths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text