Thermo Scientific Chemicals

D-Arginine monohydrochloride, 98%

Catalog number: A16222.06
5 g, Each
Thermo Scientific Chemicals

D-Arginine monohydrochloride, 98%

Catalog number: A16222.06
5 g, Each
Quantity
Catalog number: A16222.06
also known as A16222-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
627-75-8
IUPAC Name
hydrogen (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Molecular Formula
C6H15ClN4O2
InChI Key
KWTQSFXGGICVPE-PGMHMLKASA-N
SMILES
[H+].[Cl-].N[C@H](CCCN=C(N)N)C(O)=O
Form
Powder
Optical Rotation
-22? ? 1.5? (c=2 in 6M HCl)
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream

Description

The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

Solubility
Soluble in water (50 mg/ml), and 3N HCl (5).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text