Thermo Scientific Chemicals

Canada balsam, natural filtered, Thermo Scientific Chemicals

Catalog number: A16289.36
500 g, Each
Thermo Scientific Chemicals

Canada balsam, natural filtered, Thermo Scientific Chemicals

Catalog number: A16289.36
500 g, Each
Quantity
Catalog number: A16289.36
also known as A16289-36
Price (USD)
Price: 416.00
Online price: 353.65
Your price:
Quantity
-

Chemical Identifiers

CAS
8007-47-4
Appearance (Color)
Clear colorless to yellow to green or pale pink to pale brown
Identification (FTIR)
Conforms
Refractive Index
1.5200-1.5310 @ 20°C
Form
Viscous liquid

Description

Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

Solubility
It is soluble in benzene, chloroform, xylene, ethyl acetate, ether, oil of cedar, and oil of turpentine. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text