Thermo Scientific Chemicals

Nonafluoro-1-iodobutane, 97%, Thermo Scientific Chemicals

Catalog number: A16671.14
25 g, Each
Thermo Scientific Chemicals

Nonafluoro-1-iodobutane, 97%, Thermo Scientific Chemicals

Catalog number: A16671.14
25 g, Each
Quantity
Catalog number: A16671.14
also known as A16671-14
Price (USD)
Price: 60.10
Online price: 51.65
Your price:
Quantity
-

Chemical Identifiers

CAS
423-39-2
IUPAC Name
1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular Formula
C4F9I
InChI Key
PGRFXXCKHGIFSV-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Appearance (Color)
Clear colorless or pink to purple
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3270-1.3330 @ 20°C
Identification (FTIR)
Conforms

Description

Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark away from oxidizing agents. Incompatible with alkali metals and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text