Thermo Scientific Chemicals

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A16752.14
25 g, Each
Thermo Scientific Chemicals

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A16752.14
25 g, Each
Quantity
Catalog number: A16752.14
also known as A16752-14
Price (USD)
Price: 208.00
Online price: 180.65
Your price:
Quantity
-

Chemical Identifiers

CAS
578-66-5
IUPAC Name
quinolin-8-amine
Molecular Formula
C9H8N2
InChI Key
WREVVZMUNPAPOV-UHFFFAOYSA-N
SMILES
NC1=C2N=CC=CC2=CC=C1
Melting Point (clear melt)
62.0-68.0?C
Appearance (Color)
Cream to yellow to brown
Form
Crystals or powder or crystalline powder or liquid as melt
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text