Thermo Scientific Chemicals

5-Hydroxy-1-tetralone, 99%, Thermo Scientific Chemicals

Catalog number: A17034.06
5 g, Each
Thermo Scientific Chemicals

5-Hydroxy-1-tetralone, 99%, Thermo Scientific Chemicals

Catalog number: A17034.06
5 g, Each
Quantity
Catalog number: A17034.06
also known as A17034-06
Price (USD)
Price: 166.00
Online price: 143.65
Your price:
Quantity
-

Chemical Identifiers

CAS
28315-93-7
IUPAC Name
5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
Molecular Formula
C10H10O2
InChI Key
YPPZCRZRQHFRBH-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C1CCCC2=O
Appearance (Color)
Brown to cream to pink
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
205.5-211.5?C
Assay (GC)
≥98.5%

Description

5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

Solubility
Soluble in alcohol, ether, benzene and acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text