Thermo Scientific Chemicals

DL-Arginine, 98%

Catalog number: A17520.14
25 g, Each
Thermo Scientific Chemicals

DL-Arginine, 98%

Catalog number: A17520.14
25 g, Each
Quantity
Catalog number: A17520.14
also known as A17520-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7200-25-1
IUPAC Name
(2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Formula
C6H14N4O2
InChI Key
ODKSFYDXXFIFQN-BYPYZUCNSA-N
SMILES
N[C@@H](CCCN=C(N)N)C(O)=O
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤2.5%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text