Thermo Scientific Chemicals

Tetraethyl orthocarbonate, 97+%, Thermo Scientific Chemicals

Catalog number: A17835.14
25 g, Each
Thermo Scientific Chemicals

Tetraethyl orthocarbonate, 97+%, Thermo Scientific Chemicals

Catalog number: A17835.14
25 g, Each
Quantity
Catalog number: A17835.14
also known as A17835-14
Price (USD)
Price: 163.00
Online price: 138.65
Your price:
Quantity
-

Chemical Identifiers

CAS
78-09-1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.3905-1.3945 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.

Solubility
Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol.

Notes
Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert atmosphere and refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text