Thermo Scientific Chemicals

Di-n-octylamine, 96%, Thermo Scientific Chemicals

Catalog number: A18017.22
100 g, Each
Thermo Scientific Chemicals

Di-n-octylamine, 96%, Thermo Scientific Chemicals

Catalog number: A18017.22
100 g, Each
Quantity
Catalog number: A18017.22
also known as A18017-22
Price (USD)
Price: 88.80
Online price: 75.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1120-48-5
IUPAC Name
dioctylamine
Molecular Formula
C16H35N
InChI Key
LAWOZCWGWDVVSG-UHFFFAOYSA-N
SMILES
CCCCCCCCNCCCCCCCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.4415-1.4455 @ 20?C

Description

Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

Solubility
Immiscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text