2-Benzoylnaphthalin, 98 %, Thermo Scientific Chemicals
2-Benzoylnaphthalin, 98 %, Thermo Scientific Chemicals
2-Benzoylnaphthalin, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Benzoylnaphthalin, 98 %, Thermo Scientific Chemicals

CAS: 644-13-3 | C17H12O | 232.28 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer A18121.03
auch als A18121-03 bezeichnet
Preis (EUR)
46,40
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
46,40
Each
Chemikalien-Kennzeichnungen
CAS644-13-3
IUPAC Name(naphthalen-2-yl)(phenyl)methanone
Molecular FormulaC17H12O
InChI KeySJNXJRVDSTZUFB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Melting Point (clear melt)77-85?C
Appearance (Color)White to pale yellow or pale cream
Assay (GC)≥97.5%
FormPowder
2-Benzoylnaphthalene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
2-Benzoylnaphthalin wird als pharmazeutisches Zwischenprodukt verwendet.

Löslichkeit
Löslich in heißem Methanol.

Hinweise
Stabil unter empfohlenen Lagerbedingungen. Nicht kompatibel mit Oxidationsmitteln.
RUO – Research Use Only

Allgemeine Referenzen:

  1. D.Bhattacharya; S.Sharma; A.P.Singh. Selective benzoylation of naphthalene to 2-benzoylnaphthalene using zeolite H-beta catalysts. Applied Catalysis A: General. 1997, 150,(1), 53-62.
  2. John R.Ebdon; Ian Soutar; Paul Brown; Anthony R.Lane; Alasdair J.McCabe; Linda Swanson. Luminescence studies of polymer matrices: 2. on the phosphorescence characteristics of 2-benzoylnaphthalene dispersed in various acrylic polymers. Hochleistungspolymere. 1999, 11,(1), 49-62.