Thermo Scientific Chemicals

Diphenylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A18265.22
100 g, Each
Thermo Scientific Chemicals

Diphenylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A18265.22
100 g, Each
Quantity
Catalog number: A18265.22
also known as A18265-22
Price (USD)

Chemical Identifiers

CAS
122-39-4
IUPAC Name
N-phenylaniline
Molecular Formula
C12H11N
InChI Key
DMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILES
N(C1=CC=CC=C1)C1=CC=CC=C1
Form
Flakes
Identification (FTIR)
Conforms
Appearance (Color)
White to yellow to pale brown
Melting Point (clear melt)
52-55?C
Assay (GC)
≥98.0%

Description

Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

Solubility
Soluble in ethanol, acetone, benzene, carbon tetrachloride, pyridine and ethyl acetate. Slightly soluble in chloroform.

Notes
Incompatible with strong acids, strong oxidizing agents. Keep the container tightly closed in a cool and well-ventilated area.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text