Thermo Scientific Chemicals

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Catalog number: A18288.22
100 g, Each
Thermo Scientific Chemicals

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Catalog number: A18288.22
100 g, Each
Quantity

Chemical Identifiers

CAS
306-94-5
IUPAC Name
octadecafluoro-decahydronaphthalene
Molecular Formula
C10F18
InChI Key
UWEYRJFJVCLAGH-UHFFFAOYSA-N
SMILES
FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F
Assay (GC)
≥94.0% (sum of isomers)
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3115-1.3175 @ 20?C

Description

Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text