Field's Färbemittel B
Field's Färbemittel B
Field's Färbemittel B
Thermo Scientific Chemicals

Field's Färbemittel B

CAS: 17372-87-1 | C20H6Br4Na2O5 | 691.86 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10g
50g
Katalognummer A18578.18
auch als A18578-18 bezeichnet
Preis (EUR)
81,30
Each
Menge:
50g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
81,30
Each
Chemikalien-Kennzeichnungen
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Pink to red/dark red
lambda-max (UV-Vis)(0.1% w/v in water) : 510-525nm
Specific Absorbance (E 1%/1cm)350-400 (@ lambda max)
Solution Test(0.1% w/v in water) : Clear solution
Loss on Drying≤8.0% (110°C/1hr)
Mehr anzeigen
Used to stain blood smears to identify malarial parasites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only