Thermo Scientific Chemicals

Sodium tin(IV) oxide trihydrate, 96%, Thermo Scientific Chemicals

Catalog number: A18611.36
500 g, Each
Thermo Scientific Chemicals

Sodium tin(IV) oxide trihydrate, 96%, Thermo Scientific Chemicals

Catalog number: A18611.36
500 g, Each
Quantity
Catalog number: A18611.36
also known as A18611-36
Price (USD)
Price: 114.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12209-98-2
IUPAC Name
disodium oxostannanebis(olate) trihydrate
Molecular Formula
H6Na2O6Sn
InChI Key
SFXJSNATBHJIDS-UHFFFAOYSA-N
SMILES
O.O.O.[Na+].[Na+].[O-][Sn]([O-])=O
Assay from Supplier's CofA
Total Tin, Sn: ≥42.5% (Iodometric Titration)
Residual water
≤2.0% (Gravimetry)
Impurity content
Sodium Hydroxide, NaOH: ≤1.5% (Acidimetry)
Form
Crystals or powder or crystalline powder
Assay (special)
≥95.5% (based on total Tin, Sn)

Description

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text