Gibco™ PSC Definitive Endoderm Induction Kit consists of two xeno-free media that enable efficient induction of human pluripotent stem cells자세히 알아보기
Have Questions?
카탈로그 번호
수량
A3062601
1 kit
카탈로그 번호 A3062601
제품 가격(KRW)
1,247,000
온라인 행사
Ends: 30-Sep-2026
1,385,000
할인액 138,000 (10%)
Each
수량:
1 kit
제품 가격(KRW)
1,247,000
온라인 행사
Ends: 30-Sep-2026
1,385,000
할인액 138,000 (10%)
Each
Gibco™ PSC Definitive Endoderm Induction Kit consists of two xeno-free media that enable efficient induction of human pluripotent stem cells (PSCs) to definitive endoderm. Unlike other methods that require multiple components and take five or more days, the PSC Definitive Endoderm Induction Kit can generate ≥90% CXCR4+/PDGFRα- definitive endoderm cells with only two components in just two days. Each medium is supplied as a 1X complete medium, requiring no mixing in of additional components, and the resultant definitive endoderm is capable of differentiating into midgut/hindgut, pancreatic endoderm, or liver bud progenitors that express relevant physiological markers.
Generation of high quality definitive endoderm Definitive endoderm generated using the PSC Definitive Endoderm Induction Kit is capable of differentiating into downstream lineages such as midgut/hindgut (expressing CDX2), pancreatic endoderm (expressing PDX2), and liver bud progenitors (expressing AFP).
Rapid production of definitive endoderm The PSC Definitive Endoderm Induction Kit is capable of producing definitive endoderm in only two days.
Generation of highly pure definitive endoderm The PSC Definitive Endoderm Induction Kit is capable of producing highly pure populations of definitive endoderm from pluripotent stem cells, showing ≥90% CXCR4+/PDGFRα- across multiple lines and high expression of key markers SOX17 and FOXA2.
No mixing in of components required Each medium is supplied at a 1X concentration, requiring no mixing in of reagents. Simply thaw the media and add to PSC culture.
사양
세포 유형Stem Cells (Human)
수량1 kit
배송 조건Dry Ice
제품 유형PSC Definitive Endoderm Induction Kit
Unit SizeEach
구성 및 보관
• PSC Definitive Endoderm Induction Medium A • PSC Definitive Endoderm Induction Medium B
자주 묻는 질문(FAQ)
Can I cryopreserve definitive endoderm cells?
We don't recommend cryopreserving cells at the definitive endoderm stage. After definitive endoderm induction is complete, we recommend using the cells either for characterization or immediately starting downstream differentiation.
Can these cells be maintained in definitive endoderm medium?
No. Once definitive endoderm cells are induced, we recommend using the cells either for characterization or immediately starting downstream differentiation.
Definitive endoderm cells generated using the PSC Definitive Endoderm Induction kit can be characterized by FACS or immunocytochemistry by looking at high expression of endoderm markers, including CXCR4, SOX17, and FOXA2.
I see floating cells after addition of medium B when using the PSC Definitive Endoderm Induction Kit. Is that normal?
It is normal to see floating cells after addition of medium B. Floating cells are not detrimental to definitive endoderm efficiency. Higher confluency (>15-30%) at the time of induction will generally lead to higher number of floating cells.
Generation of human iPS cell line CBTCi001-A from dermal fibroblasts obtained from a healthy donor.
Authors:Martins GLS, Paredes BD, Sampaio GLA, Nonaka CKV, da Silva KN, Allahdadi KJ, França LSA, Soares MBP, Dos Santos RR, Souza BSF
Journal:Stem Cell Res
PubMed ID:31706097
Human-induced pluripotent stem cell (hiPSC) CBTCi001-A line was generated from a healthy 30-year old male dermal fibroblasts using non-integrative reprogramming method using episomal-based plasmids expressing OCT4, SOX2, KLF4, and MYCL. Characterization of CBTCi001-A was confirmed by the expression of typical markers of pluripotency and differentiation potential in vitro. ... More
Ultrasensitive and rapid quantification of rare tumorigenic stem cells in hPSC-derived cardiomyocyte populations.
Authors:Wang Z, Gagliardi M, Mohamadi RM, Ahmed SU, Labib M, Zhang L, Popescu S, Zhou Y, Sargent EH, Keller GM, Kelley SO
Journal:Sci Adv
PubMed ID:32440533
The ability to detect rare human pluripotent stem cells (hPSCs) in differentiated populations is critical for safeguarding the clinical translation of cell therapy, as these undifferentiated cells have the capacity to form teratomas in vivo. The detection of hPSCs must be performed using an approach compatible with traceable manufacturing of ... More