Pierce™ Intact Protein Standard Mix
Thermo Scientific™

Pierce™ Intact Protein Standard Mix

The Thermo Scientific Pierce Intact Protein Standard Mix is a high quality mixture of recombinant proteins for use as aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A335275 x 76 μg
A335261 x 76 μg
Catalog number A33527
Price (USD)
836.65
Online exclusive
890.00
Save 53.35
Each
Quantity:
5 x 76 μg
Price (USD)
836.65
Online exclusive
890.00
Save 53.35
Each
The Thermo Scientific Pierce Intact Protein Standard Mix is a high quality mixture of recombinant proteins for use as a quality control standard for LC and LC/MS, LC/MS method development, and intact protein mass measurements.

• Verified protein sequences—protein sequences confirmed by MS with the average and monoisotopic masses corresponding to each provided sequence
• Recombinant—proteins expressed in E. coli and B. subtilis; no bovine sources
• Compatible—protein mixture designed for QC over a wide range of molecular weights (10-70 kDa) and m/z distribution (600-1500 m/z)
• Flexible—for use in LC, LC-MS, and direct infusion methods
• Stable—provided in a stable lyophilized format

The Pierce Intact Protein Standard Mix contains a mixture of six highly pure recombinant proteins, including human IGF-1 LR3, human thioredoxin, Streptococcus dysgalactiae Protein G, Bovine Carbonic Anhydrase II, Streptococcus Protein AG (chimeric), and Escherichia coli Exo Klenow, expressed in bacteria. These proteins were selected based on their wide range of molecular weights, m/z distribution, and MS fragmentation patterns. After expression and purification, proteins are optimally mixed and lyophilized for stability and convenience. Each vial contains 76 μg of lyophilized protein mixture and does not contain salts or detergents.

Applications:
• Qualitative LC
• LC/MS standardization and method development
• LC/MS intact protein mass measurement and top-down method development
• Digestion protocol optimization

Protein sequences for Pierce Intact Protein Mix components (FASTA format) ›

Related products
Pierce Peptide Retention Time Calibration Mixture
Pierce 6 Protein Digest, equimolar, LC-MS grade
Pierce HeLa Protein Digest Standard

For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mass Spectrometer
Quantity5 x 76 μg
Shipping ConditionWet Ice
Workflow StepCalibration
FormPowder
Product LinePierce
Product TypeIntact Protein Standard Mix
Unit SizeEach
Contents & Storage
5 vials, store at -5 to -30°C

Frequently asked questions (FAQs)

Which standards and calibrants do you recommend using for assessing LC-MS system suitability?

Here are our recommendations:

- Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321) for LC system and gradient optimization
- Pierce BSA Protein Digest, MS grade (Cat. No. 88341), Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342), and Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) for "bottom up" LC-MS method optimization and LC-MS suitability assessment
- Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) (Cat. No. A40010) for assessing the gradient, sensitivity, and dynamic range of the LC-MS system for discovery and targeted peptide quantitative workflows
- Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for assessing system performance for TMT-based quantitative workflows
- Pierce Intact Protein Standard Mix (Cat. Nos. A33526, A33527) for "top down" LC-MS method optimization and LC-MS suitability assessment

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.