L-Cysteine HCl H<sub>2</sub>O
Ajinomoto Group

L-Cysteine HCl H2O

L-cysteine hydrochloride monohydrate is a stable, crystalline amino acid derivative that serves as a nutritional supplement, food additive, and biochemical reagent.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMengeVerpackung
A341501 kgPoly Drum
A3415310 kgPoly Drum
A3415420 kgFiber Drum
A2130220 kgFiber Box
Katalognummer A34150
Preis (EUR)
-
Menge:
1 kg
Verpackung:
Poly Drum

L-cysteine hydrochloride monohydrate is a stable, crystalline amino acid derivative that serves as a nutritional supplement, food additive, and biochemical reagent. This compound represents the hydrochloride salt form of L-cysteine, which confers enhanced stability and solubility characteristics compared to the free amino acid form. Ajinomoto Health & Nutrition North America, Inc., produces this high-quality product as part of their commitment to advancing nutrition and health applications through amino acid technologies with primary applications in the biotechnology and pharmaceutical sectors.

Complies with United States Pharmacopeia (USP) and European Pharmacopoeia (EP) standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7048-04-6
Chemischer Name oder MaterialL-Cysteine hydrochloride monohydrate
VerpackungPoly Drum
GüteEP/USP
Molekulargewicht (g/mol)175.63
Reinheit (%)98.5 to 101.0%
Menge1 kg
Unit SizeEach