Product Image
Ion Torrent™

Oncomine™ Focus Assay, AmpliSeq™ Library

The Oncomine Focus Assay is a targeted next-generation sequencing (NGS), multi-biomarker assay that enables the detection of variants across 52Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A3595716 Samples
A2923048 Samples
Catalog number A35957
Price (USD)
7,520.00
Each
Quantity:
16 Samples
Price (USD)
7,520.00
Each

The Oncomine Focus Assay is a targeted next-generation sequencing (NGS), multi-biomarker assay that enables the detection of variants across 52 cancer-relevant genes from DNA and RNA in a single workflow. The assay provides reagents for library construction and two pools - one DNA and one RNA - of multiplex PCR primers for preparation of amplicon libraries from formalin-fixed paraffin embedded (FFPE) tumor samples. Using as little as 10 ng of nucleic acid per primer pool, the assay enables analysis of hotpots, SNVs, indels, CNVs, and gene fusions in a single sequencing run.

The Oncomine Focus Assay, AmpliSeq Library, is for use as part of the Ion Chef and Ion GeneStudio S5 Systems workflow. Scalability and flexibility are achieved using Ion Xpress or IonCode barcode adapters for multiplexing barcoded samples on Ion S5 chips. For a kit that also includes reagents and consumables for automated library preparation on the Ion Chef System, please see the Oncomine Focus Assay, Chef-Ready Library.

From sample to answer

The Oncomine Focus Library Assay is part of an NGS workflow that helps researchers filter variant results from hundreds to just a few key cancer driver variants. Enabled by Oncomine informatics, this filtering, as well as the annotations from on-market labels, established guidelines (US-NCCN, US-FDA, EMSO, EMA), and global clinical trials, helps you contextualize and interpret findings to move toward the answers you need.

Learn more about Oncomine Focus Assay and Oncomine oncology.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chip CompatibilityIon 520, Ion 530
For Use With (Application)Oncology Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of SamplesUp to 16 Samples
Nucleic Acid TypeDNA and RNA
Product LineOncomine
Product TypeAssay
Quantity16 Samples
Sample TypeFFPE Tissue, RNA
Shipping ConditionDry Ice
ThroughputUp to 8 Samples per Ion 520 Chip
Workflow StepPanel and Library
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

How many samples can I process with the Oncomine Focus Assay, Chef-Ready Library?

The Oncomine Focus Assay, Chef-Ready Library contains the following components needed to prepare libraries for 32 samples:

- Oncomine Focus Assay, DNA Chef-Ready Panel (2X) - contains the DNA primer pool
- Oncomine Focus Assay, RNA Chef-Ready Panel (2X) - contains the RNA primer pool
- Ion AmpliSeq Kit for Chef Reagents DL8 cartridge - allows automation of the library prep on the Ion Chef Instrument

Note: The SuperScript transcriptase for cDNA synthesis is not included and must be purchased separately.