Aliphatic Amine Latex Beads, 2% w/v, 3 μm
Invitrogen™

Aliphatic Amine Latex Beads, 2% w/v, 3 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from자세히 알아보기
Have Questions?
카탈로그 번호수량
A3736815 mL
카탈로그 번호 A37368
제품 가격(KRW)
794,000
온라인 행사
Ends: 30-Jun-2026
934,000
할인액 140,000 (15%)
Each
수량:
15 mL
제품 가격(KRW)
794,000
온라인 행사
Ends: 30-Jun-2026
934,000
할인액 140,000 (15%)
Each
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aliphatic amine latex has a high density of amine groups attached to the terminus of an aliphatic six-carbon spacer arm. This product can be used to covalently couple proteins. Location of the amine at the end of the spacer arm minimizes difficulties with steric hindrance and conformation since the group has a greater freedom to rotate. In addition, spacer arms improve the kinetics of latex agglutination reactions. The particles are stabilized by the positively-charged amine groups under low to neutral pH conditions. Care should be taken not to use them under high pH.

Microspheres Are Available in Many Sizes and Surface Functionalities
25 sizes to choose from—0.02 microns to 16 microns
20 different surface—for ready conjugation to virtually any material
Surfactant free—ultraclean
Various quantities—from milliliter to 500-liter amounts

Key Applications of Microspheres
Instrument calibration (flow cytometry, microscopy, HTS, HCS)
Flow testing (microfluidics, blood flow, water flow, and air flow)
Cell biology tracers (cell differentiation and cell tracing)
Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
사양
제품라인IDC
수량15 mL
배송 조건Room Temperature
표면 수정Aliphatic Amine
직경(미터법)3 μm
물질Polystyrene
제품 유형Latex Beads
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C).