Pierce™ Protease Inhibitor XL Capsules, EDTA-free
Thermo Scientific™
Pierce™ Protease Inhibitor XL Capsules, EDTA-free
Thermo Scientific Pierce Protease Inhibitor XL Capsules, EDTA-free, contain a broad spectrum formulation of four protease inhibitors and each capsuleRead more
Have Questions?
Catalog NumberQuantity
A3798910 Capsules
Catalog number A37989
Price (TWD)
38,850.00
Online offer
Ends: 31-Dec-2026
55,500.00
Save 16,650.00 (30%)
Each
Quantity:
10 Capsules
Price (TWD)
38,850.00
Online offer
Ends: 31-Dec-2026
55,500.00
Save 16,650.00 (30%)
Each
Thermo Scientific Pierce Protease Inhibitor XL Capsules, EDTA-free, contain a broad spectrum formulation of four protease inhibitors and each capsule is sufficient for 500 mL of solution.
Features of EDTA-free Protease Inhibitor XL Capsules:
• Broad spectrum formulation—contains AEBSF, bestatin, E-64, and pepstatin A for better protection versus PMSF
• Large volume format—one capsule is sufficient for 500 mL of solution
• Convenient—ready-to-use, dissolves clearly into solution within three minutes
• Compatible—use directly with Pierce protein assays
• EDTA-free—formulated without EDTA, a metalloproteinase inhibitor
Pierce Protease Inhibitor XL Capsules protect proteins from proteolytic degradation through inhibition of the serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. The tablets contain AEBSF, bestatin, E-64, and pepstatin A. Each capsule is sufficient for 500 mL of solution, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids. Proteases reside in cellular compartments, but upon cell lysis the proteases are released, resulting in the degradation of valuable proteins. For this reason, it is common practice to add multiple compounds known to inactivate or inhibit proteases to cell lysis buffers or cell extracts. PMSF is commonly used for protease protection in large volumes, but does not offer the same level of protection as the capsule formulation.
To use Pierce Protease Inhibitor XL Capsules, simply twist the capsule to open and empty the contents of one capsule into 500 mL of solution. The protease inhibitor formulation is compatible with most detergent-based cell lysis reagents, including Pierce cell lysis solutions. Pierce Protease Inhibitor XL Capsules do not contain phosphatase inhibitors. If necessary, phosphatase inhibitors, such as Pierce phosphatase inhibitors, can be added along with the protease inhibitor capsules.
Related products
Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
Halt Phosphatase Inhibitor Cocktail, EDTA-free (100X)
T-PER Tissue Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
InhibitorsProtease Inhibitor
Quantity10 Capsules
Shipping ConditionRoom Temperature
Sufficient For500 mL Solution
FormSolid
Product LinePierce
Product TypeProtease Inhibitor Cocktail
Unit SizeEach
Contents & Storage
1 Vial, Store at 2°C to 8°C
Frequently asked questions (FAQs)
Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?
We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What formats of protease inhibitors do you offer?
We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
I am doing large-scale cell/tissue lysis in my lab. Do you have a convenient form of protease inhibitors that I can use?
Yes, our Pierce Protease Inhibitor XL Capsules, EDTA-Free (Cat. No. A37989) are designed for this purpose. Each capsule is sufficient for 500 mL of solution.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.